5-Bromo-2-methylimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Warning
OR18106-250MG OR18106-100MG OR18106-1G
APOSOR18106-250MGEA 132 EUR
APOSOR18106-250MG APOSOR18106-100MG APOSOR18106-1G
5-Bromo-2-methylimidazo[1,2-a]pyridine
5-Bromo-2-methylimidazo[1,2-a]pyridine
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699211
CAS Number: 74420-51-2