5-Methyl-1-propyl-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
Warning
OR18119-1G OR18119-5G OR18119-10G
APOSOR18119-1GEA 55.1 EUR
APOSOR18119-1G APOSOR18119-5G APOSOR18119-10G
5-Methyl-1-propyl-1H-pyrazole-4-carbaldehyde
5-Methyl-1-propyl-1H-pyrazole-4-carbaldehyde
Formula: C₈H₁₂N₂O
MW: 152.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02855988
CAS Number: 890652-02-5