6-Bromo-o-toluidine
Supplier: Apollo Scientific
Warning
OR18267-25G
APOSOR18267-25GEA 92.8 EUR
APOSOR18267-25G
6-Bromo-o-toluidine
6-Bromo-o-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 105…107 °C/205mmHg
Density: 1.478 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD04039885
CAS Number: 53848-17-2