N-Butylaniline 99%
Supplier: Apollo Scientific
Warning
OR18275-100G OR18275-50G OR18275-500G
APOSOR18275-100GEA 29 EUR
APOSOR18275-100G APOSOR18275-50G APOSOR18275-500G
N-Butylaniline 99%
N-Butylaniline
Formula: (C₆H₅)NHCH₂CH₂CH₂CH₃
MW: 149.24 g/mol
Boiling Pt: 236…242 °C
Melting Pt: –14.4 °C
Density: 0.932 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00035798
CAS Number: 1126-78-9
EINECS: 214-425-6
UN: 2738
ADR: 6.1,II