2,6-Di-tert-butyl-4-methoxyphenol 97%
Supplier: Apollo Scientific
Warning
OR18279-100G OR18279-25G
APOSOR18279-100GEA 154 EUR
APOSOR18279-100G APOSOR18279-25G
2,6-Di-tert-butyl-4-methoxyphenol 97%
2,6-Di-tert-butyl-4-methoxyphenol
Formula: C₁₅H₂₄O₂
Boiling Pt: 135…140 °C/10mmHg
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00008824
CAS Number: 489-01-0
EINECS: 207-693-0