[2.2]-Paracyclophane 98%
Supplier: Apollo Scientific
Warning
Synonyms: Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
OR18296-25G OR18296-5G OR18296-100G
APOSOR18296-25GEA 40.6 EUR
APOSOR18296-25G APOSOR18296-5G APOSOR18296-100G
[2.2]-Paracyclophane 98%
[2.2]-Paracyclophane
Formula: C₁₆H₁₆
MW: 208.3 g/mol
Melting Pt: 283…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00003707
CAS Number: 1633-22-3
EINECS: 216-644-2