2,5-Dibromo-1,4-dimethoxybenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene
OR18298-25G OR18298-5G OR18298-10G
APOSOR18298-25GEA 88.6 EUR
APOSOR18298-25G APOSOR18298-5G APOSOR18298-10G
2,5-Dibromo-1,4-dimethoxybenzene 97%
2,5-Dibromo-1,4-dimethoxybenzene
Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00025694
CAS Number: 2674-34-2