N,N,N',N'-Tetramethyl-1,3-propanediamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: N,N,N',N'-Tetramethyl-1,3-diaminopropane, TMPDA, N1,N1,N3,N3-Tetramethylpropane-1,3-diamine, 1,3-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyltrimethylenediamine
OR18301-100G OR18301-25G OR18301-500G
APOSOR18301-100GEA 24.8 EUR
APOSOR18301-100G APOSOR18301-25G APOSOR18301-500G
N,N,N',N'-Tetramethyl-1,3-propanediamine 98%
N,N,N',N'-Tetramethyl-1,3-propanediamine
Formula: C₇H₁₈N₂
MW: 130.23 g/mol
Boiling Pt: 145…146 °C
Melting Pt: –70 °C
Density: 0.779 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00008337
CAS Number: 110-95-2
EINECS: 203-818-8
UN: 2734
ADR: 8+3,II