Ethylvanillin 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde
OR18302-500G OR18302-100G
APOSOR18302-500GEA 79.9 EUR
APOSOR18302-500G APOSOR18302-100G
Ethylvanillin 99%
Ethylvanillin
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 285 °C
Melting Pt: 74…79 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890