2-Chlorobenzothiazole 98%
Supplier: Apollo Scientific
Warning
OR18309-100G OR18309-25G
APOSOR18309-100GEA 46.4 EUR
APOSOR18309-100G APOSOR18309-25G
2-Chlorobenzothiazole 98%
2-Chlorobenzothiazole
Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C/30mmHg
Melting Pt: 21…23 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005776
CAS Number: 615-20-3