1,2,3-Triazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole
OR18314-25G OR18314-100G
APOSOR18314-25GEA 21.9 EUR
APOSOR18314-25G APOSOR18314-100G
1,2,3-Triazole 98%
1,2,3-Triazole
Formula: C₂H₃N₃
Boiling Pt: 203 °C/752mmHg
Melting Pt: 23…25 °C
Density: 1.192 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014490
CAS Number: 288-36-8