2-Amino-4-methylpyrimidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methylpyrimidin-2-ylamine, 4-Methylpyrimidin-2-amine
OR18316-100G
APOSOR18316-100GEA 108 EUR
APOSOR18316-100G
2-Amino-4-methylpyrimidine 98%
2-Amino-4-methylpyrimidine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006101
CAS Number: 108-52-1
EINECS: 203-591-5