4-Methoxybenzyl alcohol 98%
Supplier: Apollo Scientific
Danger
Synonyms: Anisyl alcohol, p-Anisyl Alcohol, 4-(Hydroxymethyl)anisole, Anise alcohol, (4-Methoxyphenyl)methanol
OR18325-100G OR18325-1KG OR18325-500G
APOSOR18325-100GEA 21.9 EUR
APOSOR18325-100G APOSOR18325-1KG APOSOR18325-500G
4-Methoxybenzyl alcohol 98%
4-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 258…260 °C
Melting Pt: 22…25 °C
Density: 1.108 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6