1,1'-Carbonyldiimidazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
OR18334-1KG OR18334-250G OR18334-50G
APOSOR18334-1KGEA 128 EUR
APOSOR18334-1KG APOSOR18334-250G APOSOR18334-50G
1,1'-Carbonyldiimidazole 98%
1,1'-Carbonyldiimidazole
Formula: C₇H₆N₄O
MW: 162.15 g/mol
Melting Pt: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8,II