3-(3,4-Dimethoxyphenyl)propionic acid 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid
OR18337-100G OR18337-25G
APOSOR18337-100GEA 69.6 EUR
APOSOR18337-100G APOSOR18337-25G
3-(3,4-Dimethoxyphenyl)propionic acid 99%
3-(3,4-Dimethoxyphenyl)propionic acid
Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002774
CAS Number: 2107-70-2
EINECS: 218-288-3