Cyclopropylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: Aminocyclopropane
OR18344-100G OR18344-250G
APOSOR18344-100GEA 48 EUR
APOSOR18344-100G APOSOR18344-250G
Cyclopropylamine 98%
Cyclopropylamine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 49…50 °C
Melting Pt: –10 °C
Density: 0.824 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
UN: 2733
ADR: 3+8,II