2,3-Dimethyl-2-butene
Supplier: Apollo Scientific
Danger
Synonyms: Tetramethylethylene
OR18350-100G OR18350-25G
APOSOR18350-100GEA 0 EUR
APOSOR18350-100G APOSOR18350-25G
2,3-Dimethyl-2-butene
2,3-Dimethyl-2-butene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 72…73 °C
Melting Pt: –75 °C
Density: 0.708 g/cm³ (25 °C)
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008897
CAS Number: 563-79-1
EINECS: 209-263-8
UN: 3295
ADR: 3,II