5-Chloroisatin
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-1H-indole-2,3-dione, 5-Chloroindoline-2,3-dione
OR18353-100G OR18353-25G
APOSOR18353-100GEA 58 EUR
APOSOR18353-100G APOSOR18353-25G
5-Chloroisatin
5-Chloroisatin
Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
Melting Pt: 254…258 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014567
CAS Number: 17630-76-1
EINECS: 241-614-0