3-Phenoxybenzyl alcohol
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Hydroxymethyl)diphenyl ether, (3-Phenoxyphenyl)methanol
OR18354-25G OR18354-100G OR18354-500G
APOSOR18354-25GEA 21.9 EUR
APOSOR18354-25G APOSOR18354-100G APOSOR18354-500G
3-Phenoxybenzyl alcohol
3-Phenoxybenzyl alcohol
Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
Boiling Pt: 135…140 °C/0.1mmHg
Melting Pt: 44381 °C
Density: 1.149 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004636
CAS Number: 13826-35-2
EINECS: 237-525-1
UN: 3082
ADR: 9+EHS III