4-Chlorobenzophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: p-Chlorobenzophenone
OR18380-1KG OR18380-500G OR18380-5KG
APOSOR18380-1KGEA 48 EUR
APOSOR18380-1KG APOSOR18380-500G APOSOR18380-5KG
4-Chlorobenzophenone 97%
4-Chlorobenzophenone
Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 330…332 °C
Melting Pt: 74…77 °C
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00000622
CAS Number: 134-85-0
EINECS: 205-160-7