2-Chloro-5-iodobenzoic acid 98%
Supplier: Apollo Scientific
Danger
OR18410-100G OR18410-25G
APOSOR18410-100GEA 45.1 EUR
APOSOR18410-100G APOSOR18410-25G
2-Chloro-5-iodobenzoic acid 98%
2-Chloro-5-iodobenzoic acid
Formula: C₇H₄ClIO₂
MW: 282.46 g/mol
Melting Pt: 156…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1 III