2-Amino-N-methylbenzamide
Supplier: Apollo Scientific
Warning
OR1843-25G OR1843-5G OR1843-100G
APOSOR1843-25GEA 89.9 EUR
APOSOR1843-25G APOSOR1843-5G APOSOR1843-100G
2-Amino-N-methylbenzamide
2-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Melting Pt: 75…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060602
CAS Number: 4141-08-6