(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%
Supplier: Apollo Scientific
Warning
Synonyms: (1S,2R)-(-)-cis-1-Amino-2-indanol, (1S,2R)-(-)-cis-1-Aminoindan-2-ol
OR18545-100G OR18545-5G OR18545-25G
APOSOR18545-100GEA 166 EUR
APOSOR18545-100G APOSOR18545-5G APOSOR18545-25G
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 99%
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00216655
CAS Number: 126456-43-7