2-Ethynylpyridine 98%
Supplier: Apollo Scientific
Warning
OR18606-25G OR18606-5G OR18606-10G
APOSOR18606-25GEA 200 EUR
APOSOR18606-25G APOSOR18606-5G APOSOR18606-10G
2-Ethynylpyridine 98%
2-Ethynylpyridine
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 77…78 °C/14mmHg
Density: 0.94 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041598
CAS Number: 1945-84-2