4,4-Dimethylcyclohexanone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4,4-Dimethylcyclohexan-1-one
OR18607-100G OR18607-25G OR18607-5G
APOSOR18607-100GEA 253 EUR
APOSOR18607-100G APOSOR18607-25G APOSOR18607-5G
4,4-Dimethylcyclohexanone 98%
4,4-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Melting Pt: 41…45 °C
Density: 0.893 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00234965
CAS Number: 4255-62-3
UN: 1325
ADR: 4.1,III