3-Bromoisoxazole-5-carboxylic acid 97%
Supplier: Apollo Scientific
Warning
OR18624-1G OR18624-5G
APOSOR18624-1GEA 62.5 EUR
APOSOR18624-1G APOSOR18624-5G
3-Bromoisoxazole-5-carboxylic acid 97%
3-Bromoisoxazole-5-carboxylic acid
Formula: C4H2BrNO3
MW: 191.9676 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD07368611
CAS Number: 6567-35-7