1-Chloro-2,4-dinitrobenzene 98%
Supplier: Apollo Scientific
|
Danger
|
OR1867-100G
OR1867-2.5KG
OR1867-500G
APOSOR1867-100GEA
42.2
EUR
APOSOR1867-100G
APOSOR1867-2.5KG
APOSOR1867-500G
1-Chloro-2,4-dinitrobenzene 98%
1-Chloro-2,4-dinitrobenzene
Formula:
C₆H₃ClN₂O₄ MW: 202.55 g/mol Boiling Pt: 315 °C Melting Pt: 50…54 °C Density: 1.498 g/cm³ (25 °C) Flash Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007075 CAS Number: 97-00-7 EINECS: 202-551-4 UN: 3441 ADR: 6.1,II |