2-Amino-5-bromonicotinic acid 95%

Supplier: Apollo Scientific
Warning

OR1871-1G OR1871-5G
APOSOR1871-1GEA 21.9 EUR
APOSOR1871-1G APOSOR1871-5G
2-Amino-5-bromonicotinic acid 95%
2-Amino-5-bromonicotinic acid
Formula: C₆H₅BrN₂O₂
Melting Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD01860227
CAS Number: 52833-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR