Toluene-4-sulphonamide 98%
Supplier: Apollo Scientific
Warning
Synonyms: PTSA, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide
OR18741-500G OR18741-100G OR18741-1KG
APOSOR18741-500GEA 65.4 EUR
APOSOR18741-500G APOSOR18741-100G APOSOR18741-1KG
Toluene-4-sulphonamide 98%
Toluene-4-sulphonamide
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 221 °C/10mmHg
Melting Pt: 136…140 °C
Flash Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00011692
CAS Number: 70-55-3
EINECS: 200-741-1