2-Aminothiazole 97%

Supplier: Apollo Scientific
Warning

APOSOR1880-1KGEA 162 EUR
APOSOR1880-1KG
2-Aminothiazole 97%
2-Aminothiazole
Formula: C₃H₄N₂S
MW: 100,14 g/mol
Boiling Pt: 117 °C/15mmHg
Melting Pt: 88…93 °C
Storage Temperature: Refrigerator
EINECS: 202-511-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR