2,2,2-Tribromoethanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: Tribromoethyl alcohol
OR1882-25G OR1882-100G OR1882-5G
APOSOR1882-25GEA 111 EUR
APOSOR1882-25G APOSOR1882-100G APOSOR1882-5G
2,2,2-Tribromoethanol 97%
2,2,2-Tribromoethanol
Formula: Br₃CCH₂OH
Boiling Pt: 92…93 °C/10mmHg
Melting Pt: 73…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1