N-Fmoc-L-alanine 95%
Supplier: Apollo Scientific
Warning
Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH
OR18835-100G OR18835-500G OR18835-1KG
APOSOR18835-100GEA 46.4 EUR
APOSOR18835-100G APOSOR18835-500G APOSOR18835-1KG
N-Fmoc-L-alanine 95%
N-Fmoc-L-alanine
Formula: C₁₈H₁₇NO₄
Melting Pt: 147…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037139
CAS Number: 35661-39-3
EINECS: 252-660-6