1-Nitronaphthalene 99%
Supplier: Apollo Scientific
Danger
Synonyms: α-Nitronaphthalene
OR1884-100G OR1884-500G
APOSOR1884-100GEA 29 EUR
APOSOR1884-100G APOSOR1884-500G
1-Nitronaphthalene 99%
1-Nitronaphthalene
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 303…305 °C
Melting Pt: 53…58 °C
Density: 1.223 g/cm³ (25 °C)
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003913
CAS Number: 86-57-7
EINECS: 201-684-5
UN: 2538
ADR: 4.1,III