4'-Hydroxy-2'-methylacetophenone
Supplier: Apollo Scientific
Synonyms: 4′-Hydroxy-2′-methylacetophenone, 1-(4-Hydroxy-2-methylphenyl)ethanone
OR1891-100G OR1891-25G
APOSOR1891-100GEA 509 EUR
APOSOR1891-100G APOSOR1891-25G
4'-Hydroxy-2'-methylacetophenone
4'-Hydroxy-2'-methylacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002303
CAS Number: 875-59-2