1-Chloro-2-iodobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-1-iodobenzene
OR1893-100G OR1893-25G OR1893-500G
APOSOR1893-100GEA 43.5 EUR
APOSOR1893-100G APOSOR1893-25G APOSOR1893-500G
1-Chloro-2-iodobenzene 97%
1-Chloro-2-iodobenzene
Formula: C₆H₄ClI
Boiling Pt: 234…235 °C
Melting Pt: 1 °C
Density: 1.952 g/cm³ (25 °C)
Flash Pt: 111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001033
CAS Number: 615-41-8
EINECS: 210-423-4