3-(2-Bromophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR19-10G OR19-1G
APOSOR19-10GEA 314 EUR
APOSOR19-10G APOSOR19-1G
3-(2-Bromophenyl)-1H-pyrazole
3-(2-Bromophenyl)-1H-pyrazole
Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01940435
CAS Number: 114382-20-6