4-Nitrophenylhydrazine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Nitrophenyl)hydrazine hydrochloride, (4-Nitrophenyl)hydrazine hydrochloride, 4-Nitrophenylhydrazinium chloride
OR1938-100G OR1938-25G OR1938-5G
APOSOR1938-100GEA 168 EUR
APOSOR1938-100G APOSOR1938-25G APOSOR1938-5G
4-Nitrophenylhydrazine hydrochloride
4-Nitrophenylhydrazine hydrochloride
Formula: C₆H₇N₃O₂·HCl
MW: 189.6 g/mol
Melting Pt: 198…200 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00040592
CAS Number: 636-99-7
UN: 2811
ADR: 6.1,III