1,1'-Bis(diphenylphosphino)ferrocene 99%
Supplier: Apollo Scientific
Warning
Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene
OR1945-100G OR1945-25G OR1945-5G
APOSOR1945-100GEA 79.9 EUR
APOSOR1945-100G APOSOR1945-25G APOSOR1945-5G
1,1'-Bis(diphenylphosphino)ferrocene 99%
1,1'-Bis(diphenylphosphino)ferrocene
Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
Melting Pt: 181…183 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001422
CAS Number: 12150-46-8
EINECS: 430-420-3