3,5-Dibromopyridine 98%
Supplier: Apollo Scientific
Warning
OR1947-100G OR1947-25G OR1947-500G
APOSOR1947-100GEA 49.3 EUR
APOSOR1947-100G APOSOR1947-25G APOSOR1947-500G
3,5-Dibromopyridine 98%
3,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 222 °C
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4