2-Ethoxybenzoic acid 98+%
Supplier: Apollo Scientific
Warning
OR19479-500G
APOSOR19479-500GEA 133 EUR
APOSOR19479-500G
2-Ethoxybenzoic acid 98+%
2-Ethoxybenzoic acid
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 174…176 °C/15mmHg
Melting Pt: 19…20 °C
Density: 1.105 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002438
CAS Number: 134-11-2
EINECS: 205-130-3