Cyanomethylenetributylphosphorane 94%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Tributylphosphoranylidene)acetonitrile, {[Tri(but-1-yl)]phosphoranylidene}acetonitrile
OR19490-1G OR19490-5G
APOSOR19490-1GEA 172 EUR
APOSOR19490-1G APOSOR19490-5G
Cyanomethylenetributylphosphorane 94%
Cyanomethylenetributylphosphorane
Formula: C₁₄H₂₈NP
MW: 241.36 g/mol
Boiling Pt: 359.8 °C
Density: 0.921 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321151
CAS Number: 157141-27-0
UN: 2811
ADR: 6.1,III