cis,cis-9,12-Octadecadien-1-ol 95%
Supplier: Apollo Scientific
Warning
Synonyms: (9Z,12Z)-Octadeca-9,12-dien-1-ol, Linoleyl Alcohol, cis,cis-9,12-Octadecadienyl-1-ol
OR19496-5ML OR19496-1ML
APOSOR19496-5MLEA 207 EUR
APOSOR19496-5ML APOSOR19496-1ML
cis,cis-9,12-Octadecadien-1-ol 95%
cis,cis-9,12-Octadecadien-1-ol
Formula: C₁₈H₃₄O
MW: 266.47 g/mol
Boiling Pt: 155 °C/0.5mmHg
Density: 0.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00056667
CAS Number: 506-43-4
EINECS: 208-038-1