3-Butyne-1-ol
Supplier: Apollo Scientific
Warning
Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne
OR1952-25G OR1952-100G OR1952-500G
APOSOR1952-25GEA 56.2 EUR
APOSOR1952-25G APOSOR1952-100G APOSOR1952-500G
3-Butyne-1-ol
3-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 128…130 °C
Melting Pt: –63.6 °C
Density: 0.926 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
EINECS: 213-161-9
UN: 1986
ADR: 3+6.1 III