1-Bromo-3,5-dichlorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dichlorobromobenzene
OR19527-100G OR19527-250G OR19527-25G OR19527-1KG
APOSOR19527-100GEA 45.1 EUR
APOSOR19527-100G APOSOR19527-250G APOSOR19527-25G APOSOR19527-1KG
1-Bromo-3,5-dichlorobenzene
1-Bromo-3,5-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 232 °C
Melting Pt: 73…77 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7