6-Nitro-1,3-benzodioxol-5-amine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-6-nitro-1,3-benzodioxole
OR19538-1G OR19538-500MG OR19538-5G
APOSOR19538-1GEA 68.3 EUR
APOSOR19538-1G APOSOR19538-500MG APOSOR19538-5G
6-Nitro-1,3-benzodioxol-5-amine
6-Nitro-1,3-benzodioxol-5-amine
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00184722
CAS Number: 64993-07-3