3,4-Xylidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3,4-Dimethylaniline, 4-Amino-o-xylene
OR1954-1KG OR1954-250G
APOSOR1954-1KGEA 306 EUR
APOSOR1954-1KG APOSOR1954-250G
3,4-Xylidine 97%
3,4-Xylidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 226…228 °C
Melting Pt: 48…52 °C
Density: 1.076 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007810
CAS Number: 95-64-7
EINECS: 202-437-4
UN: 3452
ADR: 6.1,II