5-Nitro-1,10-phenanthroline 98%
Supplier: Apollo Scientific
Warning
OR19583-25G OR19583-5G
APOSOR19583-25GEA 198 EUR
APOSOR19583-25G APOSOR19583-5G
5-Nitro-1,10-phenanthroline 98%
5-Nitro-1,10-phenanthroline
Formula: C₁₂H₇N₃O₂
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS Number: 4199-88-6