1-Bromo-4-chlorobenzene 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromochlorobenzene
OR1960-1KG OR1960-250G OR1960-5KG
APOSOR1960-1KGEA 55.1 EUR
APOSOR1960-1KG APOSOR1960-250G APOSOR1960-5KG
1-Bromo-4-chlorobenzene 98+%
1-Bromo-4-chlorobenzene
Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C
Melting Pt: 63…67 °C
Density: 1.651 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3