4-(1-Pyrrolidinyl)pyridine
Supplier: Apollo Scientific
Danger
Synonyms: 4-Pyrrolidinopyridine
OR1965-100G OR1965-25G OR1965-5G
APOSOR1965-100GEA 132 EUR
APOSOR1965-100G APOSOR1965-25G APOSOR1965-5G
4-(1-Pyrrolidinyl)pyridine
4-(1-Pyrrolidinyl)pyridine
Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00006415
CAS Number: 2456-81-7
UN: 2923
ADR: 8+6.1 III