1,8-Diazabicyclo[5.4.0]undec-7-ene
Supplier: Apollo Scientific
Danger
Synonyms: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU
OR1967-250G OR1967-500G OR1967-1KG
APOSOR1967-250GEA 55.1 EUR
APOSOR1967-250G APOSOR1967-500G APOSOR1967-1KG
1,8-Diazabicyclo[5.4.0]undec-7-ene
1,8-Diazabicyclo[5.4.0]undec-7-ene
Formula: C₉H₁₆N₂
MW: 152.24 g/mol
Boiling Pt: 80…83 °C
Melting Pt: –70 °C
Density: 1.018 g/cm³ (25 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006930
CAS Number: 6674-22-2
EINECS: 229-713-7
UN: 3267
ADR: 8,II